Assay Details
Gene Symbol: ENPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018863.3 | XP_004018912.2 | 3 - 4 | 1116 | 100 | |
XM_012100761.2 | XP_011956151.2 | 3 - 4 | 1167 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
