Assay Details
Gene Symbol: BCL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956472.1 | XP_027812273.1 | 4 - 5 | 499 | 103 | |
XM_027956473.1 | XP_027812274.1 | 4 - 5 | 499 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
