Assay Details
Gene Symbol: LOC114115233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970740.1 | XP_027826541.1 | 2 - 3 | 250 | 91 |
Gene Symbol: LOC101102485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970981.1 | XP_027826782.1 | 9 - 10 | 844 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
