Assay Details
Gene Symbol: LOC114115233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970740.1 | XP_027826541.1 | 6 - 7 | 819 | 68 |
Gene Symbol: LOC101102485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970981.1 | XP_027826782.1 | 13 - 14 | 1413 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
