Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020820.4 | XP_004020869.3 | 18 - 19 | 2585 | 61 | |
XM_012104209.2 | XP_011959599.2 | 18 - 19 | 2447 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
