Assay Details
Gene Symbol: RETREG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012940.4 | XP_004012989.1 | 1 - 2 | 363 | 68 | |
XM_012186125.3 | XP_012041515.1 | 1 - 2 | 373 | 68 | ||
XM_027974489.1 | XP_027830290.1 | 1 - 2 | 368 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
