Assay Details
Gene Symbol: LOC101103602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001935.4 | XP_004001984.1 | 10 - 11 | 1365 | 64 | |
XM_012130794.3 | XP_011986184.1 | 10 - 11 | 1365 | 64 | ||
XM_012131126.3 | XP_011986516.1 | 6 - 7 | 851 | 64 | ||
XM_012131595.3 | XP_011986985.1 | 5 - 6 | 697 | 64 | ||
XM_027969579.1 | XP_027825380.1 | 10 - 11 | 1402 | 64 | ||
XM_027969591.1 | XP_027825392.1 | 10 - 11 | 1401 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
