Assay Details
Gene Symbol: LOC101103602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001935.4 | XP_004001984.1 | 13 - 14 | 1559 | 68 | |
XM_012131207.3 | XP_011986597.1 | 12 - 13 | 1539 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
