Assay Details
Gene Symbol: LOC101103916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100463.2 | XP_014955949.1 | 3 - 4 | 329 | 87 | |
XM_015100464.2 | XP_014955950.1 | 2 - 3 | 170 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
