Assay Details
Gene Symbol: ADGRG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100451.2 | XP_014955937.2 | 5 - 6 | 539 | 58 | |
XM_027977874.1 | XP_027833675.1 | 2 - 3 | 535 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
