Assay Details
Gene Symbol: DMAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015271.3 | XP_004015320.2 | 1 - 2 | 60 | 82 | |
XM_027978348.1 | XP_027834149.1 | 1 - 2 | 60 | 82 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DMAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015271.3 | XP_004015320.2 | 1 - 2 | 60 | 82 | |
XM_027978348.1 | XP_027834149.1 | 1 - 2 | 60 | 82 |