Assay Details
Gene Symbol: IL3RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963322.1 | XP_027819123.1 | 6 - 7 | 763 | 103 | |
XM_027963323.1 | XP_027819124.1 | 6 - 7 | 1021 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
