Assay Details
Gene Symbol: SETD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102043.2 | XP_014957529.2 | 8 - 9 | 2023 | 63 | |
XM_027957380.1 | XP_027813181.1 | 8 - 9 | 1023 | 63 | ||
XM_027957381.1 | XP_027813182.1 | 9 - 10 | 1007 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
