Assay Details
Gene Symbol: UBL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963034.1 | XP_027818835.1 | 2 - 3 | 226 | 105 | |
XM_027963035.1 | XP_027818836.1 | 2 - 3 | 228 | 105 |

More Information
Set Membership: |
![]() ![]() |
