Assay Details
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019650.4 | XP_004019699.1 | 2 - 3 | 308 | 95 | |
XM_012102095.3 | XP_011957485.1 | 2 - 3 | 378 | 95 | ||
XM_012102096.3 | XP_011957486.1 | 2 - 3 | 341 | 95 | ||
GenBank mRNA | KY320499.1 | - | 2 - 3 | 272 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
