Assay Details
Gene Symbol: WDR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102056.2 | XP_014957542.2 | 1 - 2 | 313 | 103 | |
XM_027957418.1 | XP_027813219.1 | 1 - 2 | 263 | 103 | ||
XM_027957419.1 | XP_027813220.1 | 1 - 2 | 240 | 103 | ||
XM_027957420.1 | XP_027813221.1 | 1 - 2 | 311 | 103 | ||
XM_027957421.1 | XP_027813222.1 | 1 - 2 | 311 | 103 | ||
XM_027957422.1 | XP_027813223.1 | 1 - 2 | 310 | 103 | ||
XM_027957424.1 | XP_027813225.1 | 1 - 2 | 314 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027957418.1, XM_027957419.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
