Assay Details
Gene Symbol: MACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007779.4 | XP_004007828.1 | 3 - 4 | 2315 | 86 | |
XM_012176466.3 | XP_012031856.1 | 4 - 5 | 2416 | 86 | ||
XM_027968502.1 | XP_027824303.1 | 3 - 4 | 3040 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
