Assay Details
Gene Symbol: LOC101105780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101638.2 | XP_014957124.2 | 2 - 3 | 563 | 88 | |
XM_027956516.1 | XP_027812317.1 | 1 - 2 | 1148 | 88 | ||
XM_027956517.1 | XP_027812318.1 | 1 - 2 | 1150 | 88 | ||
XM_027956518.1 | XP_027812319.1 | 2 - 3 | 490 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
