Assay Details
Gene Symbol: ZC3HAV1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095401.2 | XP_014950887.2 | 1 - 2 | 1280 | 159 |

More Information
Set Membership: |
![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZC3HAV1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095401.2 | XP_014950887.2 | 1 - 2 | 1280 | 159 |
Set Membership: |
![]() ![]() |