Assay Details
Gene Symbol: CRYBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972398.1 | XP_027828199.1 | 12 - 13 | 5189 | 141 | |
XM_027972399.1 | XP_027828200.1 | 11 - 12 | 5101 | 141 | ||
XM_027972400.1 | XP_027828201.1 | 11 - 12 | 4902 | 141 |

More Information
Set Membership: |
![]() ![]() |
