Assay Details
Gene Symbol: PGBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959040.1 | XP_027814841.1 | 5 - 6 | 1623 | 110 | |
XM_027959041.1 | XP_027814842.1 | 5 - 6 | 864 | 110 | ||
XM_027959042.1 | XP_027814843.1 | 5 - 6 | 1176 | 110 | ||
XM_027959043.1 | XP_027814844.1 | 4 - 5 | 1905 | 110 | ||
XM_027959044.1 | XP_027814845.1 | 4 - 5 | 1061 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
