Assay Details
Gene Symbol: TMBIM7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012176316.3 | XP_012031706.1 | 4 - 5 | 529 | 79 | |
XM_012176317.3 | XP_012031707.1 | 4 - 5 | 529 | 79 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMBIM7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012176316.3 | XP_012031706.1 | 4 - 5 | 529 | 79 | |
XM_012176317.3 | XP_012031707.1 | 4 - 5 | 529 | 79 |