Assay Details
Gene Symbol: ACRV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019520.4 | XP_004019569.1 | 1 - 2 | 54 | 67 | |
XM_027959716.1 | XP_027815517.1 | 1 - 2 | 54 | 67 | ||
XM_027959717.1 | XP_027815518.1 | 1 - 2 | 54 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
