Assay Details
Gene Symbol: LOC101111528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965195.1 | XP_027820996.1 | 1 - 2 | 721 | 70 | |
XM_027965196.1 | XP_027820997.1 | 1 - 2 | 732 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
