Assay Details
Gene Symbol: LOC101111528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965195.1 | XP_027820996.1 | 6 - 7 | 1644 | 102 | |
XM_027965196.1 | XP_027820997.1 | 6 - 7 | 1655 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
