Assay Details
Gene Symbol: LOC101118793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972290.1 | XP_027828091.1 | 19 - 20 | 2882 | 116 | |
XM_027972291.1 | XP_027828092.1 | 18 - 19 | 2861 | 116 | ||
XM_027972292.1 | XP_027828093.1 | 12 - 13 | 2209 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
