Assay Details
Gene Symbol: RPL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015420.3 | XP_004015469.2 | 4 - 5 | 440 | 105 | |
XM_004015421.3 | XP_004015470.1 | 4 - 5 | 655 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
