Assay Details
Gene Symbol: SETD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956265.1 | XP_027812066.1 | 4 - 5 | 1723 | 82 | |
GenBank mRNA | JN598905.1 | - | 1 - 2 | 156 | 82 |
Gene Symbol: LOC114116628 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973704.1 | XP_027829505.1 | 1 | 414 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
