Assay Details
Gene Symbol: CDK2AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956427.1 | XP_027812228.1 | 2 - 3 | 410 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
