Assay Details
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017115.3 | XP_004017164.2 | 3 - 4 | 732 | 83 | |
XM_012096791.2 | XP_011952181.2 | 3 - 4 | 732 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
