Assay Details
Gene Symbol: BICRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978541.1 | XP_027834342.1 | 4 - 5 | 313 | 72 | |
XM_027978542.1 | XP_027834343.1 | 4 - 5 | 394 | 72 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BICRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978541.1 | XP_027834342.1 | 4 - 5 | 313 | 72 | |
XM_027978542.1 | XP_027834343.1 | 4 - 5 | 394 | 72 |