Assay Details
Gene Symbol: FAM234A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961965.1 | XP_027817766.1 | 4 - 5 | 1602 | 87 | |
XM_027961966.1 | XP_027817767.1 | 5 - 6 | 1699 | 87 | ||
XM_027961967.1 | XP_027817768.1 | 5 - 6 | 876 | 87 | ||
XM_027961968.1 | XP_027817769.1 | 5 - 6 | 724 | 87 | ||
XM_027961969.1 | XP_027817770.1 | 4 - 5 | 711 | 87 | ||
XM_027961970.1 | XP_027817771.1 | 4 - 5 | 1621 | 87 | ||
XM_027961971.1 | XP_027817772.1 | 4 - 5 | 668 | 87 | ||
XM_027961972.1 | XP_027817773.1 | 4 - 5 | 799 | 87 | ||
XM_027961973.1 | XP_027817774.1 | 4 - 5 | 777 | 87 | ||
XM_027961974.1 | XP_027817775.1 | 5 - 6 | 1697 | 87 | ||
XM_027961975.1 | XP_027817776.1 | 5 - 6 | 746 | 87 | ||
XM_027961976.1 | XP_027817777.1 | 4 - 5 | 694 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
