Assay Details
Gene Symbol: ABHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017743.4 | XP_004017792.1 | 5 - 6 | 714 | 61 |
Gene Symbol: LOC114112206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963748.1 | XP_027819549.1 | 2 - 3 | 1288 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
