Assay Details
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106260.3 | XP_011961650.1 | 3 - 4 | 988 | 96 | |
XM_027963637.1 | XP_027819438.1 | 3 - 4 | 988 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
