Assay Details
Gene Symbol: B3GAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097680.2 | XP_014953166.2 | 1 - 2 | 591 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B3GAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097680.2 | XP_014953166.2 | 1 - 2 | 591 | 72 |
Set Membership: |
![]() ![]() ![]() |