Assay Details
Gene Symbol: HDGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976536.1 | XP_027832337.1 | 2 - 3 | 483 | 82 | |
XM_027976543.1 | XP_027832344.1 | 2 - 3 | 258 | 82 | ||
XM_027976545.1 | XP_027832346.1 | 2 - 3 | 101 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
