Assay Details
Gene Symbol: PLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970123.1 | XP_027825924.1 | 5 - 6 | 790 | 89 | |
XM_027970124.1 | XP_027825925.1 | 5 - 6 | 751 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
