Assay Details
Gene Symbol: MOB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008633.4 | XP_004008682.1 | 3 - 4 | 997 | 100 | |
XM_004008634.4 | XP_004008683.1 | 3 - 4 | 890 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
