Assay Details
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098194.2 | XP_014953680.1 | 2 - 3 | 317 | 75 | |
XM_027973672.1 | XP_027829473.1 | 2 - 3 | 297 | 75 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098194.2 | XP_014953680.1 | 2 - 3 | 317 | 75 | |
XM_027973672.1 | XP_027829473.1 | 2 - 3 | 297 | 75 |