Assay Details
Gene Symbol: CCL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964274.1 | XP_027820075.1 | 2 - 3 | 3287 | 94 |
Gene Symbol: LOC114112952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964275.1 | XP_027820076.1 | 5 - 6 | 5130 | 94 | |
XM_027964276.1 | XP_027820077.1 | 5 - 6 | 5194 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
