Assay Details
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973424.1 | XP_027829225.1 | 1 - 2 | 1445 | 92 | |
XM_027973425.1 | XP_027829226.1 | 2 - 3 | 1196 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
