Assay Details
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973424.1 | XP_027829225.1 | 2 - 3 | 1615 | 111 | |
XM_027973425.1 | XP_027829226.1 | 3 - 4 | 1366 | 111 | ||
XM_027973426.1 | XP_027829227.1 | 3 - 4 | 1961 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
