Assay Details
Gene Symbol: CXADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002819.4 | XP_004002868.1 | 5 - 6 | 773 | 108 | |
XM_012187117.3 | XP_012042507.1 | 5 - 6 | 823 | 108 | ||
XM_012187131.3 | XP_012042521.1 | 5 - 6 | 773 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
