Assay Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015091887.2 | XP_014947373.2 | 2 - 3 | 603 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
