Assay Details
Gene Symbol: LOC114110793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962145.1 | XP_027817946.1 | 2 - 3 | 161 | 142 |
Gene Symbol: DOCK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092012.2 | XP_014947498.2 | 17 - 18 | 2080 | 142 | |
XM_015092013.2 | XP_014947499.2 | 17 - 18 | 2080 | 142 | ||
XM_015092014.2 | XP_014947500.2 | 17 - 18 | 2080 | 142 | ||
XM_015092015.2 | XP_014947501.2 | 17 - 18 | 2080 | 142 | ||
XM_027970750.1 | XP_027826551.1 | 17 - 18 | 2087 | 142 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027962145.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
