Assay Details
Gene Symbol: LOC114110793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962145.1 | XP_027817946.1 | 4 - 5 | 342 | 125 |
Gene Symbol: DOCK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092012.2 | XP_014947498.2 | 19 - 20 | 2261 | 125 | |
XM_015092013.2 | XP_014947499.2 | 19 - 20 | 2261 | 125 | ||
XM_015092014.2 | XP_014947500.2 | 19 - 20 | 2261 | 125 | ||
XM_015092015.2 | XP_014947501.2 | 19 - 20 | 2261 | 125 | ||
XM_027970750.1 | XP_027826551.1 | 19 - 20 | 2268 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
