Assay Details
Gene Symbol: DOHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008617.4 | XP_004008666.2 | 4 - 5 | 976 | 80 | |
XM_027969887.1 | XP_027825688.1 | 4 - 5 | 946 | 80 | ||
GenBank mRNA | EU370547.1 | - | 1 - 2 | 141 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
