Assay Details
Gene Symbol: MEAK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014942.4 | XP_004014991.2 | 5 - 6 | 1063 | 64 | |
XM_027977687.1 | XP_027833488.1 | 4 - 5 | 1061 | 64 | ||
XM_027977688.1 | XP_027833489.1 | 4 - 5 | 1020 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
