Assay Details
Gene Symbol: MEAK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014942.4 | XP_004014991.2 | 6 - 7 | 1187 | 74 | |
XM_027977687.1 | XP_027833488.1 | 5 - 6 | 1185 | 74 | ||
XM_027977688.1 | XP_027833489.1 | 5 - 6 | 1144 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
