Assay Details
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017912.4 | XP_004017961.2 | 10 - 11 | 941 | 108 | |
XM_012098847.3 | XP_011954237.2 | 16 - 17 | 2764 | 108 | ||
XM_012098849.3 | XP_011954239.2 | 15 - 16 | 2737 | 108 | ||
XM_012098850.3 | XP_011954240.2 | 16 - 17 | 2764 | 108 | ||
XM_012098851.3 | XP_011954241.2 | 15 - 16 | 2737 | 108 | ||
XM_012098852.3 | XP_011954242.2 | 11 - 12 | 890 | 108 | ||
XM_012098853.3 | XP_011954243.2 | 11 - 12 | 1958 | 108 | ||
XM_012098854.3 | XP_011954244.2 | 11 - 12 | 968 | 108 | ||
XM_012098855.3 | XP_011954245.2 | 11 - 12 | 1838 | 108 | ||
XM_012098857.3 | XP_011954247.2 | 11 - 12 | 1829 | 108 | ||
XM_012098858.3 | XP_011954248.2 | 10 - 11 | 941 | 108 | ||
XM_027957293.1 | XP_027813094.1 | 11 - 12 | 968 | 108 | ||
XM_027957294.1 | XP_027813095.1 | 11 - 12 | 1957 | 108 | ||
XM_027957295.1 | XP_027813096.1 | 11 - 12 | 1829 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
